Learn More
Electron Microscopy Sciences Aurion Tween-20 10 ML
SDP
Supplier:  Electron Microscopy Sciences 25564
Encompass_Preferred
Tween-20 is a non-ionic detergent with a molecular weight of about 600 and a critical micelle concentration (CMC) of 0.06-0.07% in water at room temperature. Its working mechanism may in part be based on its action as a detergent, binding to the hydrophobic moieties of water insoluble compounds, rendering them hydrophilic. In addition, immuno-compounds may become incorporated into micelles when the Tween-20 concentration is higher than the CMC, for instance at 0.1 % in PBS at pH 7.4.
Catalog No. 50-295-43