Learn More
Apexbio Technology LLC Aurora A Inhibitor I, 5mg. Cas: 1158838-45-9 MFCD: MFCD16495816. Aurora A inhibitor
SDP
Supplier:  Apexbio Technology LLC A41265
Encompass_Preferred
IC50: 3.4 nM.Aurora A Inhibitor I is a novel, potent, and selective inhibitor of Aurora A . Aurora kinases are reported to be required for mitosis and to complete cell division. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4886