Learn More
Selleck Chemical LLC Aurora Kinase Inhibitor III S2931-2mg
SDP
Supplier:  Selleck Chemical LLC S29312MG
Encompass_Preferred
Aurora kinase inhibitor III is a potent inhibitor of Aurora A kinase with an IC50 of 42 nM and has high selectivity for Aurora A over BMX BTK IGF-1R c-Src TRKB SYK and EGFR (IC50s 386 3 550 591 1 980 2 510 887 and 10 000 nM respectively)
Catalog No. 50-310-8698