Learn More
Cayman Chemical Aurora KInase InhIbItr II 1mg
SDP
Supplier:  Cayman Chemical 175411 MG
Encompass_Preferred
A cell-permeable anilinoquinazoline that blocks the activity of Aurora A (IC50 = 0.39 µM); blocks MCF-7 cells proliferation with an IC50 value of 1.25 µM
Catalog No. 50-296-5225