Learn More
ION OPTICKS PTY LTD Aurora® Rapid™ 8×150 XT C18 UHPLC column
Supplier:  ION OPTICKS PTY LTD AUR480150C18XT
Encompass
8 cm x 150 μm C18 UHPLC column with built-in HeatSync™ technology, compatible with EASY-Spray, Nanospray Flex and Newomics UniESI and DuoESI ion sources.
Catalog No. NC3938263