Learn More
Medchemexpress LLC Aurothiomalate sodium | 12244-57-4 | MFCD00064304 | 98.0% | 390.08 g/mol | C4H6O4S.Au.xNa | 100 MG
SDP
Supplier:  Medchemexpress LLC HY106381100MG
Encompass_Preferred
Aurothiomalate sodium is a gold-containing research compound used to study oncogenic signaling and redox biology. It functions as a PKCι signaling inhibitor and a thioredoxin reductase (TrxR) inhibitor, exhibiting anti-tumor and anti-rheumatic activity in preclinical studies. The material is supplied as a solid with 98.0% purity for laboratory use.
  • Potent PKCι signaling inhibitor used in cancer signaling studies.
  • Inhibits thioredoxin reductase and modulates NF-κB-related pathways.
  • Demonstrates anti-tumor activity in preclinical lung cancer models.
  • High purity formulation suitable for in vitro and in vivo research.
  • Provided as a stable solid for convenient handling and storage.
Catalog No. 50-002-43981