Learn More
Medchemexpress LLC HY-134640 5mg , AUTAC4 CAS:2267315-04-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1346405MG
Encompass_Preferred
HY-134640 5mg AUTAC4 CAS: 2267315-04-6 AUTAC4 is a mitochondria-targeting autophagy-targeting chimera (AUTAC). AUTAC4 downregulates cytosolic proteins and promotes targeted mitochondrial turnover via mitophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0064