Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Devices Molecular Devices Microplate Reader, Fluorescence, Dual Monochromator, 250-850 nm

Supplier:  Molecular Devices XPSG

Encompass_Preferred

The Gemini XPS microplate reader is a top-reading fluorescence spectrofluorometer that provides flexible excitation and emission selection, sensitive detection, and multiple read modes for microplate assays.

  • Dual monochromators for excitation and emission wavelength selection.
  • Excitation range 250-850 nm.
  • Emission range 360-850 nm.
  • Supports 6, 12, 24, 48, 96, and 384-well microplates.
  • Read types: endpoint, kinetic, spectral scan, and well scan.
  • Xenon flash lamp light source.
  • Photomultiplier tube detector.
  • Temperature control from ambient +4°C to 45°C.
  • Approximate weight 35 lb (16 kg) and dimensions 34 x 42 x 42 cm.

Catalog No. 50-000-14737


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.