Learn More
Sino Biological Peptide Substrates: Autocamtide 2
Supplier:  Sino Biological A1558
Encompass_Preferred
The Autocamtide 2 peptide sequence (KKALRRQETVDAL-amide) is based on the autophosphorylation site (amino acid 283-290) on CaMKII.
Catalog No. 50-256-9832