Learn More
Hoefer, Inc Multi-Channel AutoLaser Marking Machine
SDP
Supplier:  Hoefer, Inc STATL02
Encompass_Preferred
Features and Benefits:Laser-etch tri-code sample tubes with a maximum of 1200 tubes per hour, user defined with his own information. Customize the code using characters, numbers, letters, barcodes, 2D codes, graphics, etc. Laser-etched code with high contrast and readability,simply recognized with barcode reader. Open communication interface for integrating into existing software.Data can be imported externally without manual input.Compatible with the entire range of Hoefer sample storage tubes.Simplify the sample handling process by solving issues of paper labels that are prone to contamination, not waterproof, and fall off at low temperatures.
Catalog No. NC2956942
Only null left
Add to Cart
Request a Quote