Learn More
Chemglass Life Sciences Chemglass automation conductive tips, 1000 μl, for Tecan liquid handlers, 96 tips/pack (CLS-3499A-1000)
SDP
Supplier:  Chemglass Life Sciences CLS3499A1000
Encompass_Preferred
Automation conductive pipette tips designed for high-throughput liquid handling. These 1000 μl conductive tips are made from medical-grade conductive polypropylene, pre-sterilized for automated workflows, and packaged for reliable use in robotic systems.
  • 1000 μl nominal volume.
  • Made from medical-grade conductive polypropylene (black).
  • Pre-sterilized by electron beam; free of DNA, RNA, protease, and pyrogen.
  • Packed 96 tips per pack, 2,304 tips per case.
  • Designed for compatibility with Tecan automated liquid-handling systems.
  • Manufactured in a 100,000-grade clean plant with strict quality controls.
  • Separate cavity packing improves conductivity uniformity and traceability.
Catalog No. 50-273-3251