Learn More
Chemglass Life Sciences Chemglass Automation conductive filter tips, 50 μl, for Hamilton (pre-sterilized, 96/pack)
SDP
Supplier:  Chemglass Life Sciences CLS3499A052
Encompass_Preferred
Chemglass automation conductive filter tips are 50 μl, pre-sterilized conductive pipette tips designed for automated liquid-handling platforms. They provide low carryover and consistent performance in high-throughput workflows.
  • 50 μl volume per tip.
  • Medical-grade black conductive polypropylene construction.
  • Pre-sterilized by electron beam.
  • Wrapped in peelable Tyvek films and blisters for automated loading.
  • Reduces aerosol carryover and cross-contamination.
  • Uniform conductivity and smooth inner surface to minimize residual liquid.
  • Pack quantity: 96 per pack; case quantity: 2,304.
Catalog No. 50-273-3262