Learn More
Medchemexpress LLC Autophagy inducer 7 | 944159-20-0 | 410.55 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY171047100MG
Encompass_Preferred
Autophagy inducer 7 (Compound SSA) is an inducer of both autophagy and apoptosis. It works by inhibiting Akt/mTOR signaling and suppressing the expression of downstream proteins. This compound also inhibits DNA synthesis, leading to a G0-G1 cell-cycle arrest, and effectively inhibits tumor cell growth.
  • Activates autophagy by inhibiting Akt/mTOR signaling.
  • Induces apoptosis in various human lung adenocarcinoma cell lines.
  • Suppresses DNA synthesis and causes G0-G1 cell-cycle arrest.
  • Inhibits tumor cell growth.
  • Increases autophagic flux in A549 cells.
Catalog No. 50-003-86338