Learn More
Medchemexpress LLC Autophagy inducer 7 | 944159-20-0 | 410.55 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1710475MG
Encompass_Preferred
Autophagy inducer 7 (Compound SSA) is an Autophagy and Apoptosis inducer. It activates autophagy by inhibiting Akt/mTOR signaling and the expression of downstream proteins. It suppresses DNA synthesis and causes a G0-G1 cell-cycle arrest. Autophagy inducer 7 inhibits tumor cell growth.
  • Inhibits the growth of human lung adenocarcinoma cell lines
  • Induces apoptosis in various cell lines
  • Inhibits DNA synthesis and increases cells in G0-G1 phase
  • Induces autophagy and increases autophagic flux by inhibiting Akt/mTOR/p70S6k signaling
  • Induces dose- and time-dependent cleavage of cytosolic LC3-I to autophagosome-associated LC3-II
  • Decreased p62 expression
  • Displayed accumulation of vesicle-like structures within the cytoplasm
  • Increased both the number and size of acidic vesicles within the cytoplasm
Catalog No. 50-003-86332