Learn More
Enzo Life Sciences SCREEN-WELL Autophagy library (500 µl/well)
SDP
Supplier:  Enzo Life Sciences BML28370500
Encompass_Preferred
The SCREEN-WELL Autophagy library contains 94 compounds with defined autophagy-inducing or -inhibitory activity. Compounds are dissolved in DMSO or water at 10mM and aliquoted into deep-well plates at 100 or 500µl per well. A variety of structurally and mechanistically different compound classes are included. A useful tool for studying the roles of pro- and anti-autophagic molecules in cells as well as for use in in vitro applications. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0215