Learn More
Chemglass Life Sciences Autoradiography Film, 5" x 7"
SDP
Supplier:  Chemglass Life Sciences CLS190601
Encompass_Preferred
Autoradiography Film, 5" x 7". ¿ Ideal for Chemiluminescence¿ Suitable for use with 14C, 32P, 33P, 125I, 35S¿ Compatible with Automatic Processors¿ Packaged in Lined, Light-Safe, Dispenser Boxes of 100 Sheets per Box¿ High Contrast¿ Reduced Background¿ Publication Quality¿ Double EmulsionBluBlot HS is a high quality, blue sensitive film that is ideal for a wide range of electrophoretic and autoradiography applications. It can be used with calcium tungstate or blue-emitting rare-earth screens. BluBlot HS exhibits excellent sensitivity and contrast characteristics and is coated on both sides for improved consistency and overall performance. It reliably produces sharp, bright images that are easily visualized and suitable for publication.
Catalog No. 50-131-1287