Learn More
Cole-Parmer Extech EX430A True RMS autoranging digital multimeter
Supplier:  Cole-Parmer 2000534
Encompass
Precision accuracy with True RMS current/voltage at an economical price. Low current capabilitymeasure down to 0.1 A. Audible and visual input alerts. Autoranging DMM. Type K temperature. Capacitance.
Catalog No. 50-992-946