Learn More
Perkin Elmer US LLC Autosampler Add-On Kit for Clarus 690 GC
Supplier:  Perkin Elmer US LLC N6550509
Encompass_Preferred
Note: This Product Number cannot be used on Clarus 600 GC.
Catalog No. 50-210-4230
Add to Cart
Request a Quote