Learn More
Gold Biotechnology Inc Auxo Agro EHA105 Comp Cells
SDP
Supplier:  Gold Biotechnology Inc CC16810X50
Encompass_Preferred
GoldBio s Auxo-Agro competent cells are methionine auxotrophic strains of Agrobacterium which reduce overgrowth during the infection process while increasing plant transformation efficiency
Catalog No. 50-005-62261