Learn More
Medchemexpress LLC AUZ 454 | 853299-07-7 | 99.2% | 501.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15004100MG
Encompass_Preferred
AUZ 454 is a type II CDK2 inhibitor with a Kd of 8.2 nM. It inhibits CDK2 activity by competing with the binding of activating cyclins. It shows an inhibitory effect on Caki-1 and ACHN cells with WTAP overexpression and decreases colony formation through a CDK2-dependent mechanism. This product is intended for research use only.
  • Inhibits CDK2 activity
  • Competes with activating cyclins
  • Inhibits cell proliferation in Caki-1 and ACHN cells
  • Decreases colony formation in Caki-1 and ACHN cells
Catalog No. 50-003-61695