Learn More
Medchemexpress LLC Avacopan | 1346623-17-3 | MFCD28502293 | 98.5% | 581.64 g·mol⁻¹ | C33H35F4N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1762710MG
Encompass_Preferred
Avacopan (CCX168) is a selective, orally active complement 5a receptor (C5aR) inhibitor supplied as a solid research chemical. It is used as a pharmacological tool in immunology and signaling studies, with reported potency at C5aR (IC50 = 0.1 nM). Typical analytical specifications include high purity and a defined molecular weight for reproducible experimental results.
  • Selective C5aR inhibition with low-nanomolar potency.
  • High listed purity suitable for analytical and biological assays.
  • Solid powder format for convenient solution preparation.
  • Defined molecular weight for analytical confirmation.
  • Accompanied by a datasheet and certificate of analysis.
Catalog No. 50-002-98609