Learn More
Selleck Chemical LLC Avadomide (CC-122) 2mg 1015474-32-4
SDP
Supplier:  Selleck Chemical LLC S78922MG
Encompass_Preferred
Avadomide (CC-122), a new chemical entity termed pleiotropic pathway modifier, is a novel agent for Diffuse large B-cell lymphoma(DLBCL) with antitumor and immunomodulatory activity. Its molecular target is the protein cereblon (CRBN), a substrate receptor of the cullin ring E3 ubiquitin ligase complex CRL4CRBN. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5382