Learn More
TARGETMOL CHEMICALS INC AVAGACESTAT 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T62495MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Avagacestat (BMS-708163) (BMS-708163) is a potent selective orally bioavailable (gamma)-secretase inhibitor of A(beta)40 and A(beta)42 with IC50 of 0.3 nM and 0.27 nM demonstrating a 193-fold selectivity against Notch. Phase 2. purity: 99%
Catalog No. 50-257-6555