Learn More
Avanti Polar Lipids Avanti Research 20α-hydroxycholesterol, >99% Purity, powder, -20°C Storage
Supplier: Avanti Polar Lipids 700156P10MG
20α-hydroxycholesterol is a high-purity sterol available in powder form. It has been utilized in studies to investigate the impact of oxysterols on mammalian immune function.
- Purity >99% (TLC)
- Provided as a powder
- Synonyms include 5-cholestene-3α,20α-diol
- Empirical formula C27H46O2
- CAS number 516-72-3
- Molecular weight 402.65
- Recommended storage at -20°C
- Not hygroscopic or light sensitive
- Stable for 3 months
- Melting point of 125°C to 132°C
- Slightly soluble in water
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.