Learn More
Medchemexpress LLC Avasopasem manganese | 435327-40-5 | 96.1% | 483.38 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10911010MM/1ML
Encompass_Preferred
Avasopasem manganese is a potent superoxide dismutase mimetic. It rapidly and specifically converts superoxide radicals to hydrogen peroxide, thereby arresting the initiation of damaging cascades.
  • Potent superoxide dismutase mimetic
  • Rapidly and specifically converts superoxide radicals to hydrogen peroxide
  • Arrests the initiation of damaging cascades
  • Used for research of severe oral mucositis and cancer
  • Intended to interrupt severe oral mucositis pathogenesis
  • Reduces duration, incidence, and severity of radiation-induced oral mucositis
Catalog No. 50-003-45985