Learn More
Medchemexpress LLC Avasopasem manganese | 435327-40-5 | 96.1% | 483.38 g/mol | C21H35Cl2MnN5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1091105MG
Encompass_Preferred
Avasopasem manganese is a small-molecule superoxide dismutase (SOD) mimetic used in research to modulate reactive oxygen species. It rapidly converts superoxide (O2•-) to hydrogen peroxide (H2O2) and is investigated for reducing radiation-induced severe oral mucositis and for oncology research.
  • Potent superoxide dismutase mimetic
  • Rapid conversion of superoxide to hydrogen peroxide
  • Suitable for research into radiation-induced mucositis and cancer
  • Provided in small research quantities for dosing flexibility
  • High chemical purity for reproducible experimental results
Catalog No. 50-002-64410