Learn More
Selleck Chemical LLC Avatrombopag hydrochloride E4969-1G
SDP
Supplier:  Selleck Chemical LLC E49691G
Encompass_Preferred
Avatrombopag hydrochloride(AKR-501 hydrochloride YM477 hydrochloride) is an orally administered second generation agonist of Thrombopoietin receptor(TpoR) with an EC50 value of 3 3 nM It is used in the treatment of primary chronic immune thrombocytopenia (ITP)
Catalog No. 50-000-40863