Learn More
Medchemexpress LLC Avatrombopag (maleate) | 677007-74-8 | MFCD12032126 | C33H38Cl2N6O7S2 | 10MG

Supplier: Medchemexpress LLC HY13463A10MG
Avatrombopag maleate is the maleate salt of avatrombopag, an orally active thrombopoietin receptor agonist. This listing describes the analytical standard supplied as a solid for laboratory and analytical use; intended for research purposes only and not for human administration.
- Analytical standard for laboratory and analytical use.
- CAS number 677007-74-8.
- Chemical formula C33H38Cl2N6O7S2.
- Molecular weight 765.73 g/mol.
- Purity 99.9% (manufacturer reported).
- Available in small pack sizes suitable for analytical work.
- Store sealed, away from moisture; follow certificate of analysis for exact conditions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.