Learn More
Apexbio Technology LLC AVE 0991 sodium salt 306288-04-0 5mg
SDP
Supplier:  Apexbio Technology LLC B12135
Encompass_Preferred
AVE 0991 sodium salt (CAS 306288-04-0) is a synthetic non-peptide agonist of the angiotensin-(1-7) receptor Mas Acting as a functional mimic of angiotensin-(1-7) AVE 0991 competitively binds to endothelial cell membranes with an IC50 of 21 35 nM in bovine aortic endothelial cells In vitro treatment with AVE 0991 promotes nitric oxide release to levels significantly greater than angiotensin-(1-7) In vivo AVE 0991 reduces urine volume and increases urine osmolality in mice effects reversed by angiotensin II antagonists indicating receptor specificity It also inhibits atherogenesis in ApoE knockout mice supporting its application in cardiovascular and renal research
Catalog No. 50-000-87736