Learn More
Medchemexpress LLC Avenanthramide A | 299.28 g·mol-1 | C16H13NO5 | 50MG
SDP
Supplier:  Medchemexpress LLC HY11497750MG
Encompass_Preferred
Avenanthramide A is a phytoalexin which can be found in oats (Avena sativa L ) Avenanthramide A targets the RNA helicase DDX3 leads to mitochondrial swelling and increased ROS production and induces apoptosis in CRC cells Avenanthramide A exhibits antitumor efficacy in mouse model Avenanthramide A orally active[1]
Catalog No. 50-003-07564