Learn More
Medchemexpress LLC Avenanthramide C | 116764-15-9 | MFCD23380831 | 98.9% | 315.28 g/mol | C16H13NO6 | 50MG
SDP
Supplier:  Medchemexpress LLC HY12434450MG
Encompass_Preferred
Avenanthramide C is a naturally occurring avenanthramide isolated from oat (Avena sativa) and used in research for its antioxidant and anti-inflammatory properties. Supplied as a research-grade reference standard in a 50 mg solid form, it is suitable for biochemical, cellular, and analytical applications. Key chemical attributes include CAS 116764-15-9, molecular formula C16H13NO6, molecular weight 315.28 g/mol, and typical purity 98.9%.
  • High purity 98.9% as tested by supplier.
  • Supplied as a 50 mg solid for small-scale experiments.
  • Suitable for biochemical and cellular assays.
  • Known antioxidant and anti-inflammatory activity.
  • Characterized by CAS 116764-15-9 and molecular weight 315.28 g/mol.
Catalog No. 50-002-39222