Learn More
Abcam Averantin, Aflato x in B1 percursor, 1MG
Supplier: Abcam AB1442011MG
MW 372.4 Da, Purity >98%. Mycotoxin. Aflatoxin B1 precursor. Cytotoxic against solid tumor cell lines.
The product is subject to the following: Abcam Restricted Use Statement