Learn More
AdipoGen Averufin (1 mg)
SDP
Supplier:  AdipoGen AGCN20527M001
Encompass_Preferred
Chemical. CAS: 14016-29-6. Formula: C20H16O7. MW: 368.3. Isolated from fungus Aspergillus versicolor. Mycotoxin. Biosynthetic precursor of aflatoxins. Antibiotic. Shows moderate antibacterial activity against Gram-positive bacteria. Oxidative phosphorylation (OXPHOS) inhibitor. Inhibits succinate cytochrome c reductase complex (III) of the respiratory chain in mitochondria. Useful tool for immunometabolism research.
Catalog No. 50-205-3701