Learn More
Abcam AVG-Cl (Aminoetho x yvinyl glycine hydrochloride), 25MG
Supplier:  Abcam AB14538225MG
Encompass_Preferred
MW 196.63 Da, Purity >95%. Potent 1-aminocyclopropanecarboxylic acid synthase inhibitor. Inhibits cystathionine β-lyase (IC₅₀ = 32 nM). Inhibits ethylene synthesis and increases freezing tolerance in plants in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5246