Learn More
Selleck Chemical LLC Avibactam sodium 10mg 1192491-61-4 AVE-1330A, NXL104
SDP
Supplier:  Selleck Chemical LLC S373210MG
Encompass_Preferred
Avibactam (AVE-1330A, NXL104) is a covalent, reversible, non-?-lactam ?-lactamase inhibitor with IC50 values of 8, 80, and 38 nM for TEM-1, P99, and KPC-2 ?-lactamases, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8316