Learn More
Medchemexpress LLC Avibactam sodium hydrate | 2938989-90-1 | 99.6% | 305.24 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14879B10MM/1ML
Encompass_Preferred
Avibactam sodium hydrate (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor. It inhibits β-lactamase TEM-1 and CTX-M-15, as well as class A and C β-lactamases.
  • Covalent and reversible non-β-lactam β-lactamase inhibitor
  • Inhibits β-lactamase TEM-1 and CTX-M-15
  • Inhibits class A and C β-lactamases
  • Demonstrates significant therapeutic efficacy in a *K. pneumoniae* strain Y8 infected mouse model
  • Shows *in vitro* and *in vivo* bactericidal activity against Carbapenemase-producing *Klebsiella pneumoniae*
Catalog No. 50-003-61553