Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Avibactam sodium hydrate | 2938989-90-1 | 99.6% | 305.24 g/mol | C7H12N3NaO7S | 10 MG
SDP

Supplier:  Medchemexpress LLC HY14879B10MG

Encompass_Preferred

Avibactam sodium hydrate is a covalent, reversible, non-β-lactam β-lactamase inhibitor provided for research and analytical applications. It is used in enzyme inhibition assays and antibiotic resistance studies to characterize inhibition of class A and related β-lactamases.

  • Covalent and reversible non-β-lactam β-lactamase inhibitor.
  • Inhibits TEM-1 (IC50 = 8 nM) and CTX-M-15 (IC50 = 5 nM).
  • CAS number 2938989-90-1.
  • Chemical formula C7H12N3NaO7S; molecular weight 305.24 g/mol.
  • Purity 99.6%.
  • Available as solids (5 mg-1 g) and prepared solution (10 mM x 1 mL).
  • Supplied with supporting documents: data sheet, COA, SDS, H NMR, RP-HPLC.
  • Intended for research use; follow SDS for handling and storage.

Catalog No. 50-002-85644


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.