Learn More
Selleck Chemical LLC Avitinib maleate E4597-1G
SDP
Supplier:  Selleck Chemical LLC E45971G
Encompass_Preferred
Avitinib maleate(Abivertinib maleate AC0010 maleate AC0010MA) is a third-generation irreversible and orally active selective inhibitor of EGFR It displays IC50 values of 0 18 nM 0 18 nM 7 68 nM and against EGFR L858R EGFR T790M and wild-type EGFR Avitinib maleate also acts as an inhibitor of BTK that induces apoptosis of BTK in mantle cell lymphoma
Catalog No. 50-000-39850