Learn More
Medchemexpress LLC HY-16735 25mg , Avoralstat CAS:918407-35-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1673525MG
Encompass_Preferred
HY-16735 25mg Avoralstat CAS: 918407-35-9 Avoralstat (BCX4161), a potent and orally active plasma kallikrein (PKK) inhibitor, is used for hereditary angioedema research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0194