Learn More
Medchemexpress LLC Avotaciclib (trihydrochloride) | 1983984-01-5 | 99.6% | 390.66 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY137432A1MG
Encompass_Preferred
Avotaciclib trihydrochloride is an orally active cyclin-dependent kinase 1 (CDK1) inhibitor. It can inhibit the proliferation and induce apoptosis of tumor cells. This product is used in the research of cancers such as pancreatic cancer and lung cancer.
  • Inhibits cell proliferation and promotes apoptosis in radiotherapy-resistant non-small cell lung cancer cell lines.
  • Demonstrates enhanced efficacy when used in combination with certain other compounds.
  • Applicable in cancer research, including studies on pancreatic and lung cancer.
Catalog No. 50-003-51508