Learn More
New England Biolabs, Inc. AvrII – 500 units
SDP
Supplier:  New England Biolabs, Inc. R0174L
Encompass
A restriction endonuclease that recognizes the sequence C^CTAG_G.
Catalog No. 50-812-086