Learn More
Medchemexpress LLC HY-107390 5mg Medchemexpress, AX-024 CAS:1370544-73-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1073905MG
Encompass_Preferred
Medchemexpress, HY-107390 5mg AX-024 CAS:1370544-73-2 AX-024 is an orally available, first-in-class inhibitor of the TCR-Nck interaction that selectively inhibits TCR-triggered T cell activation with an IC50 ~1 nM. AX-024 modulates cell signaling by targeting SH3 domains. AX-024 has low-acute toxicity and high potency and selectivity, and strongly inhibit the production of IL-6, TNF-α, IFN-γ, IL-10 and IL-17A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8400