Learn More
TARGETMOL CHEMICALS INC AX-024 hydrochloride 10mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T440310MG
Encompass_Preferred
AX-024 hydrochloride is an cytokine release inhibitor which can strongly inhibit the production of interleukin-6 (IL-6), tumor necrosis factor-α (TNFα), interferon-γ (IFN-γ), IL-10 and IL-17A.
Catalog No. NC1624944