Learn More
TARGETMOL CHEMICALS INC AX-024 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T44035MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. AX-024 hydrochloride (AX-024 HCl) is an cytokine release inhibitor which can strongly inhibit the production of interleukin-6 (IL-6) tumor necrosis factor-(alpha) (TNF(alpha)) interferon-(gamma) (IFN-(gamma)) IL-10 and IL-17A. purity: 99%
Catalog No. 50-257-6767