Learn More
Medchemexpress LLC Axelopran sulfate | 949904-50-1 | 555.68 g/mol | C26H41N3O8S | 10 MG

Supplier: Medchemexpress LLC HY16765A10MG
Axelopran sulfate is the sulfate salt of axelopran, an opioid receptor antagonist used in research to profile μ, δ, and κ opioid receptors. It is supplied for laboratory research, is highly soluble in DMSO, and has reported pKi values of 9.8 (human μ), 8.8 (human δ), and 9.9 (guinea pig κ).
- Sulfate salt form suitable for laboratory research.
- High reported affinity for opioid receptors (pKi 9.8-9.9).
- Soluble in DMSO: ≥ 100 mg/mL.
- Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
- Intended for in vitro and preclinical opioid receptor pharmacology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.