Learn More
Medchemexpress LLC Axelopran sulfate | 949904-50-1 | 99.1% | 555.68 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY16765A5MG
Encompass_Preferred
Axelopran sulfate is an opioid receptor antagonist. It exhibits potent binding affinity with pKi values of 9.8, 8.8, and 9.9 for human recombinant μ and δ receptors and guinea pig κ receptor, respectively. This product is intended for research use only and is not sold to patients.
  • Opioid receptor antagonist
  • Potent binding affinity for μ, δ, and κ opioid receptors
  • High purity
  • Solid form
  • White to off-white appearance
  • Soluble in DMSO
Catalog No. 50-003-93544