Learn More
Medchemexpress LLC Axitinib-13C,d3 | 1261432-00-1 | 97.8% | 390.48 g·mol⁻¹ | C21H15D3N4OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10065S10MG
Encompass_Preferred
Axitinib-13C,d3 is a 13C- and deuterium-labeled isotopic standard of axitinib provided for research use. It is intended as an analytical reference for mass spectrometry, pharmacokinetic, and metabolic studies, enabling accurate quantitation and tracer experiments in preclinical and analytical workflows.
  • Isotopically labeled with 13C and d3 for use as an internal standard.
  • High isotopic enrichment (≈99% 13C, ≈98% D) for reliable mass spectrometry quantitation.
  • High purity suitable for analytical applications (~97.8% HPLC).
  • Suitable for pharmacokinetic, metabolic profiling, and tracer studies.
  • Supplied in research-scale quantities appropriate for laboratory assays.
Catalog No. 50-002-13847