Learn More
Cayman Chemical AxItInIb SulfoxIde 5mg
SDP
Supplier:  Cayman Chemical 280425 MG
Encompass_Preferred
A major inactive metabolite of axitinib; formed from axitinib primarily by CYP3A4
Catalog No. 50-293-9306