Learn More
Bioss Axl/UFO - Extracellular region
SDP
Supplier:  Bioss BSM70355M
Encompass_Preferred
Axl/UFO (Extracellular region) Antibody Primary Antibody Unconjugated Host Species Mouse Target Species Human Clonality Monoclonal Applications WB IP M047Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-65220