Learn More
Medchemexpress LLC HY-107420 5mg Medchemexpress, AY 9944 CAS:366-93-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1074205MG
Encompass_Preferred
Medchemexpress, HY-107420 5mg AY 9944 CAS:366-93-8 AY 9944 is a specific cholesterol biosynthesis inhibitor. AY 9944 inhibits the 7-dehydro cholesterol Δ7-reductase (DHCR7) enzyme (IC50=13 nM). AY 9944 causes hypocholesterolemia and accumulation of 7DHC. At high doses, AY 9944 inhibits also in cultured embryos sterol Δ7-Δ8 isomerase, which causes the accumulation of cholest-8-en-3β-ol[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8409